Merck
  • Enhanced toxicity of the protein cross-linkers divinyl sulfone and diethyl acetylenedicarboxylate in comparison to related monofunctional electrophiles.
Enhanced toxicity of the protein cross-linkers divinyl sulfone and diethyl acetylenedicarboxylate in comparison to related monofunctional electrophiles.
Chemical research in toxicology (2011-08-05)
James D West, Chelsea E Stamm, Haley A Brown, Samantha L Justice, Kevin A Morano
ZUSAMMENFASSUNG
Previously, we determined that diethyl acetylenedicarboxylate (DAD), a protein cross-linker, was significantly more toxic than analogous monofunctional electrophiles. We hypothesized that other protein cross-linkers enhance toxicity similarly. In agreement with this hypothesis, the bifunctional electrophile divinyl sulfone (DVSF) was 6-fold more toxic than ethyl vinyl sulfone (EVSF) in colorectal carcinoma cells and greater than 10-fold more toxic in Saccharomyces cerevisiae. DVSF and DAD caused oligomerization of yeast thioredoxin 2 (Trx2p) in vitro and promoted Trx2p cross-linking to other proteins in yeast at cytotoxic doses. Our results suggest that protein cross-linking is considerably more detrimental to cellular homeostasis than simple alkylation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Divinylsulfon, contains hydroquinone as inhibitor, ≥96%
Sigma-Aldrich
Dimethylacetylendicarboxylat, 95%
Sigma-Aldrich
Diethylacetylendicarboxylat, 95%