Merck
Kinetic resolution of (R,S)-2-butanol using enzymatic synthesis of esters.
Applied biochemistry and biotechnology (2011-08-13)
Maria Dolores Romero, Jose Maria Gomez, Beatriz Diaz-Suelto, Alicia Garcia-Sanz, Nicola Baster
ZUSAMMENFASSUNG
Kinetic resolution of (R,S)-2-butanol using enzymatic synthesis of esters has been studied. (R,S)-2-Butanol is commonly found as a racemic mixture, and the products of its esterification are racemic mixtures too. This work is of great significance in the field of the enzymatic kinetic resolution due to the little information found in literature about the resolution of (R,S)-2-butanol as pure compound. So, this article is a contribution about the enzymatic resolution of (R,S)-2-butanol. The reaction here studied is the esterification/transesterification of (R,S)-2-butanol in organic media (n-hexane) using as biocatalyst the lipase Novozym 435®. The main target of this study is to analyze the influence of certain variables in this reaction. Some of these variables are acyl donor (acids and esters), concentration of substrates, enzyme/substrate ratio, and temperature. The main conclusions of this study are the positive effect of higher substrates concentration (1.5 M) and larger amount of enzyme (13.8 g mol(-1) substrate) on kinetic resolution rate but not a very noticeable effect on enantiomeric excesses. The longer the carboxylic acid chain is, the better results are obtained. Besides to achieve a satisfactory kinetic resolution, it is recommendable to select reaction times (180 min) at which the highest substrate enantiomeric excess is reached (~60%). The temperature has not an appreciable influence on the resolution in the range studied (40-60 °C). When an ester (vinyl acetate) is used as acyl donor, the resolution shows better results than when using a carboxylic acid as acyl donor (ee(s) ~90% at 90 min). Moreover, Michaelis-Menten parameters, v(max) and K(M), were determined, 0.04 mol l(-1) min(-1) and 0.41 mol l(-1), respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Sigma-Aldrich
Aluminium-tri-sec-butoxid, 97%
Supelco
2-Butanol, analytical standard
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%