Merck
  • Pathway-dependent isotope fractionation during aerobic and anaerobic degradation of monochlorobenzene and 1,2,4-trichlorobenzene.
Pathway-dependent isotope fractionation during aerobic and anaerobic degradation of monochlorobenzene and 1,2,4-trichlorobenzene.
Environmental science & technology (2011-08-20)
Xiaoming Liang, Michael R Howlett, Jennifer L Nelson, Gavin Grant, Sandra Dworatzek, Georges Lacrampe-Couloume, Stephen H Zinder, Elizabeth A Edwards, Barbara Sherwood Lollar
ZUSAMMENFASSUNG
Stable carbon isotope fractionation is a valuable tool for monitoring natural attenuation and to establish the fate of groundwater contaminants. In this study, we measured carbon isotope fractionation during aerobic and anaerobic degradation of two chlorinated benzenes: monochlorobenzene (MCB) and 1,2,4-trichlorobenzene (1,2,4-TCB). MCB isotope fractionation was measured in anaerobic methanogenic microcosms, while 1,2,4-TCB isotope experiments were carried out in both aerobic and anaerobic microcosms. Large isotope fractionation was observed in both the anaerobic microcosm experiments. Enrichment factors (ε) for anaerobic reductive dechlorination of MCB and 1,2,4-TCB were -5.0‰ ± 0.2‰ and -3.0‰ ± 0.4‰, respectively. In contrast, no significant isotope fractionation was found during aerobic microbial degradation of 1,2,4-TCB. The cleavage of a C-Cl σ bond occurs during anaerobic reductive dechlorination of MCB and 1,2,4-TCB, while no σ bond cleavage is involved during aerobic degradation via dioxygenase. The difference in isotope fractionation for aerobic versus anaerobic biodegradation of MCB and 1,2,4-TCB can be explained by the difference in the initial step of aerobic versus anaerobic biodegradation pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2,4-Trichlorbenzol, suitable for HPLC, ≥99%
Sigma-Aldrich
Chlorbenzol, anhydrous, 99.8%
Sigma-Aldrich
Chlorbenzol, suitable for HPLC, 99.9%
Sigma-Aldrich
Chlorbenzol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2,4-Trichlorbenzol, ReagentPlus®, ≥99%
Sigma-Aldrich
1,2,4-Trichlorbenzol, anhydrous, ≥99%
Supelco
Chlorbenzol, analytical standard
Supelco
1,2,4-Trichlorbenzol, PESTANAL®, analytical standard