- Synthesis and biological activity of novel tiliroside derivants.
Synthesis and biological activity of novel tiliroside derivants.
European journal of medicinal chemistry (2011-08-23)
Nan Qin, Chun-Bao Li, Mei-Na Jin, Li-Huan Shi, Hong-Quan Duan, Wen-Yan Niu
PMID21856048
ZUSAMMENFASSUNG
A series of new tiliroside derivatives were synthesized and characterized by analytical (1)H NMR, (13)C NMR and mass spectrometry. All of the compounds were evaluated for anti-diabetic properties in vitro using HepG2 cells. Compounds 3c, 3d, and 3i-l caused significant enhancements in glucose consumption by insulin-resistant HepG2 cells compared with control cells and cells that were exposed to metformin (an anti-diabetic drug). Moreover, compound 3l significantly activated adenosine 5'-monophosphate-activated protein kinase activity and reduced acetyl-CoA carboxylase activity. Thus, the tiliroside derivative 3l offers potential to be developed as a new approach for treating type II diabetes.