Merck
  • Effect of cellulose acetate butyrate microencapsulated ammonium polyphosphate on the flame retardancy, mechanical, electrical, and thermal properties of intumescent flame-retardant ethylene-vinyl acetate copolymer/microencapsulated ammonium polyphosphate/polyamide-6 blends.
Effect of cellulose acetate butyrate microencapsulated ammonium polyphosphate on the flame retardancy, mechanical, electrical, and thermal properties of intumescent flame-retardant ethylene-vinyl acetate copolymer/microencapsulated ammonium polyphosphate/polyamide-6 blends.
ACS applied materials & interfaces (2011-08-24)
Bibo Wang, Qinbo Tang, Ningning Hong, Lei Song, Lei Wang, Yongqian Shi, Yuan Hu
ZUSAMMENFASSUNG
Ammonium polyphosphate (APP), a widely used intumescent flame retardant, has been microencapsulated by cellulose acetate butyrate with the aim of enhancing the water resistance of APP and the compatibility between the ethylene-vinyl acetate copolymer (EVA) matrix and APP. The structure of microencapsulated ammonium polyphosphate (MCAPP) was characterized by Fourier transform infrared spectroscopy (FTIR), X-ray photoelectron spectroscopy (XPS), scanning electron microscopy (SEM), and water contact angle (WCA). The flame retadancy and thermal stability were investigated by a limiting oxygen index (LOI) test, UL-94 test, cone calorimeter, and thermogravimetric analysis (TGA). The WCA results indicated that MCAPP has excellent water resistance and hydrophobicity. The results demonstrated that MCAPP enhanced interfacial adhesion, mechanical, electrical, and thermal stability of the EVA/MCAPP/polyamide-6 (PA-6) system. The microencapsulation not only imparted EVA/MCAPP/PA-6 with a higher LOI value and UL-94 rating but also could significantly improve the fire safety. Furthermore, the microencapsulated EVA/MCAPP/PA-6 composites can still pass the UL-94 V-0 rating after treatment with water for 3 days at 70 °C, indicating excellent water resistance. This investigation provides a promising formulation for the intumescent flame retardant EVA with excellent properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Sigma-Aldrich
Nylon 6, pellets
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Celluloseacetatbutyrat, average Mn ~70,000
Sigma-Aldrich
Celluloseacetatbutyrat
Sigma-Aldrich
Celluloseacetatbutyrat, average Mn ~30,000
Sigma-Aldrich
Celluloseacetatbutyrat, average Mn ~12,000
Sigma-Aldrich
Celluloseacetatbutyrat, average Mn ~30,000