Merck
Multi-system fibrosis and long-term use of ergotamine.
Annals of the Academy of Medicine, Singapore (2011-08-27)
Mohammad Hassan Murad, Fletcher A Miller, James Glockner
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ergotamin -D-tartrat, ≥97.0% (calc. based on dry substance, NT)