Merck
"Alternately divided" epidermal nevus of the fingers.
Pediatric dermatology (2011-09-13)
Daniele Torchia, Janelle Vega, Maria Miteva, Paolo Romanelli, Lawrence A Schachner
ZUSAMMENFASSUNG
A 2-year-old white girl with divided (or kissing) epidermal nevus of the third and fourth fingers of the left hand is described. The possible pathogenesis of this unique lesion is also discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tazarotene, ≥98% (HPLC)