Merck
  • Photosensitized xanthone-based oxidation of guanine and its repair: a laser flash photolysis study.
Photosensitized xanthone-based oxidation of guanine and its repair: a laser flash photolysis study.
Journal of photochemistry and photobiology. B, Biology (2011-09-13)
Ruizhi Tang, Peng Zhang, Haixia Li, Yancheng Liu, Wenfeng Wang
ZUSAMMENFASSUNG
The photosensitized oxidation of guanine (G) by the triplet state of xanthone (XT) and the repair for photo-damaged G(-H)(·) by ferulic acid (FCA) were investigated using the laser flash photolysis technique. The rate constants of the reaction of triplet state of XT with G and with FCA were determined as 4.5×10(9) and 8.0×10(9) L mol(-1) s(-1), respectively. Laser exposure was performed on the N(2)-saturated acetonitrile/water (v/v, 1:1) solution containing G, XT and FCA. The transient absorption spectra indicated that the triplet state of XT first reacted with G predominantly to form the oxidized radical G(-H)(·). The radical G(-H)(·) was rapidly repaired by FCA, and the rate constant for the repair reaction was determined as 1.1×10(9) L mol(-1) s(-1). These results demonstrated that non-enzymatic repair is a feasible method for repairing photosensitized DNA bases oxidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Xanthon, 97%