Merck
Spectroscopy and single-molecule emission of a fluorene-terthiophene oligomer.
The journal of physical chemistry. B (2011-09-16)
G E Khalil, A M Adawi, B Robinson, A J Cadby, W C Tsoi, J-S Kim, A Charas, J Morgado, D G Lidzey
ZUSAMMENFASSUNG
We study the thiophene-based oligomer poly[2,7-(9,9-bis(2'-ethylhexyl)fluorene)-alt-2,5-terthiophene] (PF3T) in solution and when dispersed at low concentration into a polynorbornene matrix. We find that at high concentration in solution the 0-0 electronic transition observed in fluorescence is suppressed, a result indicative of the formation of weakly coupled H-aggregates. At low concentration in a polymer matrix, emission from both single molecules and molecular aggregates is observed. We find that the fluorescence spectra of most PF3T emitters are composed of a number of relatively narrow emission features, indicating that the emission usually occurs from multiple chromophores. A small number of PF3T molecules are however characterized by single chromophore emission, spectral blinking, and narrowed emission peaks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2′:5′,2′′-Terthiophen, 99%