Merck
  • Vibrational and electronic investigations, thermodynamic parameters, HOMO and LUMO analysis on crotonaldehyde by ab initio and DFT methods.
Vibrational and electronic investigations, thermodynamic parameters, HOMO and LUMO analysis on crotonaldehyde by ab initio and DFT methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-09-21)
A Jayaprakash, V Arjunan, Sujin P Jose, S Mohan
ZUSAMMENFASSUNG
The energy, geometrical parameters and vibrational wavenumbers of crotonaldehyde were calculated by using ab initio and B3LYP with 6-31G(d,p) and 6-311G(d,p) basis sets. The FT-IR and FT-Raman spectra for liquid state crotonaldehyde have been recorded in the region 3400-400 cm(-1) and 3400-100 cm(-1), respectively and compared with the theoretical spectrographs constructed from the scaled harmonic vibrational frequencies calculated at HF and DFT levels. The difference between the observed and scaled wavenumber values of most of the fundamentals is very small. Detailed interpretations on vibrational modes have been made on the observed and theoretical spectra and PED for each mode was also reported more precisely. HOMO and LUMO energy levels are constructed and the corresponding theoretical frontier energy gaps are calculated to realise the charge transfer occurring in the molecule. The thermodynamic properties of the title compound have been calculated at different temperatures and the results reveals the standard heat capacities (C(0)(p)), standard entropies (S(0)) and standard enthalpy changes (ΔH(0)) increases with rise in temperature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Crotonaldehyd, vorwiegend trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Crotonaldehyd (cis+trans), ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)