Merck
  • Phosphine-initiated general base catalysis: facile access to benzannulated 1,3-diheteroatom five-membered rings via double-Michael reactions of allenes.
Phosphine-initiated general base catalysis: facile access to benzannulated 1,3-diheteroatom five-membered rings via double-Michael reactions of allenes.
Organic letters (2011-09-22)
Judy Szeto, Vardhineedi Sriramurthy, Ohyun Kwon
ZUSAMMENFASSUNG
General base-catalyzed double-Michael reactions of allenes with various dinucleophiles are described. The reactions are facilitated most efficiently by a catalytic amount of trimethylphosphine, affording six types of C2-functionalized benzannulated five-membered heterocycles: benzimidazolines, benzoxazolines, benzothiazolines, 1,3-benzodioxoles, 1,3-benzoxathioles, and 1,3-benzodithioles. This atom-economical reaction is operationally simple and provides the product heterocycles in good to excellent yields. Careful mechanistic studies unveiled the phosphine-triggered general base catalysis pathway. Furthermore, the double-Michael reaction can serve as an alternative method for the selective monoketalization of β-diketones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethylphosphin, 97%
Sigma-Aldrich
Trimethylphosphin -Lösung, 1.0 M in THF
Sigma-Aldrich
Trimethylphosphin -Lösung, 1.0 M in toluene