Merck
  • Anti-albuminuric effects of the angiotensin AT1 receptor blocker telmisartan in hypertensive patients.
Anti-albuminuric effects of the angiotensin AT1 receptor blocker telmisartan in hypertensive patients.
Clinical and experimental hypertension (New York, N.Y. : 1993) (2011-10-01)
José C Rodríguez-Pérez, Miguel A García-Bello, Aránzazu Anabitarte-Prieto, Osmel Companioni, Francisco J Nóvoa-Mogollón, Saturnino Suárez-Ortega, Celia Plaza-Toledano, Francisco Rodríguez-Esparragón
ZUSAMMENFASSUNG
We evaluated the anti-hypertensive and anti-albuminuric effect of the angiotensin receptor blocker telmisartan alone and in combination with torasemide and amlodipine. Patients were hypertensive, both diabetics and non-diabetics with persistent microalbuminuria. Our primary endpoint was a change in microalbuminuria levels, while the secondary endpoints were changes in systolic blood pressure (SBP), diastolic blood pressure (DBP), serum creatinine levels, and glomerular filtration rate.After the 16-week treatment period, the patients significantly reduced microalbuminuria levels (76.4 ± 52.4 μg/min; p < 0.001), SBP (16.4 ± 8.7 mmHg; p < 0.001) and DBP (17.7 ± 5.9 mmHg; p < 0.001). Both diabetics and non-diabetics showed an identical pattern of significance with respect to the whole population. Systolic blood pressure, DBP, and microalbuminuria were significantly reduced as a consequence of therapy, both in diabetics and non-diabetics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Torsemid, ≥98% (HPLC), solid
Torasemid, wasserfrei, European Pharmacopoeia (EP) Reference Standard
Torasemid für die Systemeignung, European Pharmacopoeia (EP) Reference Standard