Merck
  • Antihyperlipidemic properties of novel N-(benzoylphenyl)-5-substituted-1H-indole-2-carboxamides in Triton WR-1339-induced hyperlipidemic rats.
Antihyperlipidemic properties of novel N-(benzoylphenyl)-5-substituted-1H-indole-2-carboxamides in Triton WR-1339-induced hyperlipidemic rats.
Molecules (Basel, Switzerland) (2011-10-01)
Yusuf Al-Hiari, Ghassan Shattat, Tariq Al-Qirim, Waseem El-Huneidi, Ghassan Abu Sheikha, Suhair Hikmat
ZUSAMMENFASSUNG
In the search for new potential antihyperlipidemic agents, the present study focuses on the synthesis of novel N-(benzoylphenyl)-5-substituted-1H-indole-2-carboxamides (compounds 8-12, 15, 16, 18) and investigating their antihyperlipidemic activity using Triton WR-1339-induced hyperlipidemic rats as an experimental model. Hyperlipidemia was developed by intraperitoneal injection of Triton WR-1339 (250 mg/kg body weight). The tested animals were divided into normal control (NCG), hyperlipidemic (HG), compound 8, 9, 15, 16, 18- and bezafibrate treated groups. At a dose of 15 mg/kg body weight, compounds 9, 16, 18 and bezafibrate (100 mg/kg) significantly (p < 0.0001) reduced elevated plasma triglycerides levels after 12 h compared to the hyperlipidemic control group. However, only the group treated with compounds 9, 16 and 18 showed an obviously significant (p < 0.001) reduction in plasma total cholesterol levels after 12 h compared to the hyperlipidemic control group. Moreover, high density lipoprotein-cholesterol levels were significantly (p < 0.0001) increased in all treated groups after 12 h compared to the hyperlipidemic control group, except for compounds 8 and 15 which revealed inactive. It is therefore reasonable to assume that compounds 9, 16 and 18 may have potential in the treatment of hyperlipidemia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyloxapol, nonionic surfactant
Sigma-Aldrich
Tyloxapol, BioXtra