Merck
  • Evaluating headspace component vapor-time profiles by solid-phase microextraction with external sampling of an internal standard.
Evaluating headspace component vapor-time profiles by solid-phase microextraction with external sampling of an internal standard.
Analytical chemistry (2011-10-04)
William MacCrehan, Stephanie Moore, Michele Schantz
ZUSAMMENFASSUNG
The vapor-time profiles of explosive materials are of valuable interest to Homeland Security, providing critical information that can aid in the detection of explosive-containing devices. An approach is described that achieves reproducible characterization of volatile components as a function of time based on comparison of the sample response to an externally sampled internal standard (ESIS). Utilizing nonequilibrium solid-phase microextraction (SPME) measurements, this SPME-ESIS technique improves reproducibility (reported as percent relative standard deviation) of vapor-time profiles by approximately an order of magnitude and allows for an equitable comparison of the target compound between diverse materials. Two odorants associated with canine detection of explosives, 2-ethyl-1-hexanol and 2,4-dinitrotoluene, are used to optimize parameters for the SPME-ESIS technique.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
2,4-Dinitrotoluol, 97%
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
Titan(IV)-2-ethylhexyloxid, 95%
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)