Merck
  • Simultaneous determination of eight active components in Houttuynia cordata injection and its quality control in productive process.
Simultaneous determination of eight active components in Houttuynia cordata injection and its quality control in productive process.
Journal of separation science (2011-10-06)
Wei Ji, Kaishun Bi, Qianqian Chen, Lingyan Jiang, Ke Liang, Qing Li
ZUSAMMENFASSUNG
A simple, reliable and effective gas chromatography coupled with flame ionization detection method was developed for the simultaneous determination of eight components (α-pinene, β-pinene, myrcene, limonene, terpinen-4-ol, α-terpineol, bornyl acetate and methyl-n-nonylketone) in Chinese medicine Houttuynia cordata and its injection. The chromatographic separation of all eight components, including undecylene as internal standard was performed on a DB-1 column (30 m×0.25 mm, 0.25 μm). Excellent linear behaviors including herb and injection over the investigated concentration ranges were observed with the values of r(2) higher than 0.9990 for all analytes. Satisfactory intra-day and inter-day precisions were achieved with RSD less than 2% and the average recoveries for all analytes at three different concentrations obtained were in the range of 93.4-104.4%, with RSD ranging from 1.3 to 4.1%. The proposed method was successfully applied in the simultaneous determination of these active components in H. cordata and H. cordata injection (HCI), including the intermediate product of HCI in productive process, from different pharmaceutical factories and different production batches, indicating that the method in this paper was particularly suitable for the routine analysis of HCI and its quality control in productive process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
4-Carvomenthenol, natural, ≥95%, FG
Supelco
(+)-α-Terpineol, analytical standard
Terpinen-4-ol, primary reference standard
Sigma-Aldrich
4-Carvomenthenol, ≥95%, FCC, FG