Merck
  • Toxic impact of two organophosphate insecticides on biochemical parameters of a food fish and assessment of recovery response.
Toxic impact of two organophosphate insecticides on biochemical parameters of a food fish and assessment of recovery response.
Toxicology and industrial health (2011-10-11)
Madhusudan Reddy Narra, Ghousia Begum, Kodimyala Rajender, J Venkateswara Rao
ZUSAMMENFASSUNG
Sublethal effects of chlorpyrifos (CPF) and monocrotophos (MCP) on fish biochemical constituents were investigated along with the assessment of recovery response after cessation of intoxication. The fish, Clarias batrachus were exposed to 1.656 mg(-l) and 2.114 mg(-l) of CPF and MCP for 28 days. After 28 days, they were released in freshwater and allowed to recover for 21 days. The CPF exposure resulted in the decrease of carbohydrate and glycogen content, whereas MCP intoxication caused mixed response. Pyruvate and lactate contents were altered under the stress of CPF and MCP. Recovery of these alterations was observed after the cessation of toxicity. Exposure of C. batrachus to CPF and MCP resulted in decreased activity of lactate dehydrogenase in the kidney, liver and muscle but its activity increased in the gills. The CPF caused inhibition of succinate dehydrogenase enzyme in all tissues. Induction in the activity of malate dehydrogenase was caused by both insecticides. Glycogen phosphorylase a was induced in all tissues, whereas glycogen phosphorylase ab showed both induction and inhibition. Of the two insecticides, CPF was more toxic and the recovery response was less. These results are important in the assessment of the risk caused by organophosphate insecticides on nontarget organisms, especially the food fish.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Monocrotophos, PESTANAL®, analytical standard