- Diisobutylaluminum hydride mediated regioselective O desilylations: access to multisubstituted cyclodextrins.
Diisobutylaluminum hydride mediated regioselective O desilylations: access to multisubstituted cyclodextrins.
Angewandte Chemie (International ed. in English) (2011-10-19)
Ramprasad Ghosh, Ping Zhang, Aixia Wang, Chang-Chun Ling
PMID22006644