- Multiple state transition interface sampling of alanine dipeptide in explicit solvent.
Multiple state transition interface sampling of alanine dipeptide in explicit solvent.
The Journal of chemical physics (2011-10-21)
Wei-Na Du, Kristen A Marino, Peter G Bolhuis
PMID22010733
ZUSAMMENFASSUNG
We have applied the recently developed multiple state transition interface sampling approach to alanine dipeptide in explicit water. We extract the rate constant matrix for configurational changes between each pair of metastable states. The results are comparable with values from previous literature and show that the method is applicable to biomolecular systems.