Merck
  • Analysis of selenomethylselenocysteine and selenomethionine by LC-ESI-MS/MS with diethyl ethoxymethylenemalonate derivatization.
Analysis of selenomethylselenocysteine and selenomethionine by LC-ESI-MS/MS with diethyl ethoxymethylenemalonate derivatization.
The Analyst (2011-10-26)
Riin Rebane, Koit Herodes, Ivo Leito
ZUSAMMENFASSUNG
A simple method to identify and determine Selenomethionine (SeMet) and Selenomethylselenocysteine (Se-MeSeCys) with diethyl ethoxymethylenemalonate (DEEMM) derivatization and LC-ESI-MS/MS determination was developed. Separation of SeMet and Se-MeSeCys was achieved in 15.3 minutes. The calibration graph was linear in the range of 0.32 pmol to 49 pmol for SeMet and 0.34 pmol to 40 pmol for Se-MeSeCys. To prevent oxidation of SeMet, 2-mercaptoethanol was introduced to the calibration solutions. Detection limits were 0.1 pmol, which are comparable to LC-ICP-MS analysis. The developed method therefore offers an alternative to LC-ICP-MS offering similar sensitivity and additionally allows identification. The method was used to determine Se-MeSeCys and SeMet in onion samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethoxymethylen-malonsäure-diethylester, 99%