Merck
  • Synthesis and in vitro anticancer activity of 6,7-methylenedioxy (or 5-hydroxy-6-methoxy)-2-(substituted selenophenyl)quinolin-4-one analogs.
Synthesis and in vitro anticancer activity of 6,7-methylenedioxy (or 5-hydroxy-6-methoxy)-2-(substituted selenophenyl)quinolin-4-one analogs.
European journal of medicinal chemistry (2011-10-28)
Chien-Ting Chen, Mei-Hua Hsu, Yung-Yi Cheng, Chin-Yu Liu, Li-Chen Chou, Li-Jiau Huang, Tian-Shung Wu, Xiaoming Yang, Kuo-Hsiung Lee, Sheng-Chu Kuo
ZUSAMMENFASSUNG
6,7-Methylenedioxy (or 5-hydroxy-6-methoxy)-2-(substituted selenophenyl)quinolin-4-ones and their isosteric compounds were synthesized and evaluated for anticancer activity. Structure-activity relationships (SAR) of these compounds were established. Among all tested compounds, 6,7-methylenedioxy-2-(5-methylselenophen-2-yl)quinolin-4-one (4d) was found to be the most promising anticancer agent. In screening against NCI's 60 human tumor cell line panel, 4d exhibited highly selective and potent inhibitory activity against MDA-MB-435 melanoma. Furthermore, the results of COMPARE analysis suggested that 4d is an antimitotic agent with a different mechanism of action from the conventional antimitotic agents, such as colchicine, vinca alkaloids and paclitaxel. Therefore, 4d was identified as a new lead compound that merits further optimization.