Merck
  • Peroxymonosulfate-Co(II) oxidation system for the removal of the non-ionic surfactant Brij 35 from aqueous solution.
Peroxymonosulfate-Co(II) oxidation system for the removal of the non-ionic surfactant Brij 35 from aqueous solution.
Chemosphere (2011-11-01)
Michele Pagano, Angela Volpe, Giuseppe Mascolo, Antonio Lopez, Vito Locaputo, Ruggiero Ciannarella
ZUSAMMENFASSUNG
The non-ionic surfactant Brij 35 was effectively removed from concentrated aqueous solution by the peroxymonosulfate/Co(II) system, using oxone (2KHSO(5)·KHSO(4)·K(2)SO(4)) as a source of peroxymonosulfate. At pH=2.3 and initial Brij 35 concentration in the range 680-2410 mg L(-1), 86-94% removal was achieved after 24 h, using Co(II)=15 μM and oxone=5.9 mM. The effectiveness of removal did not change when initial pH was in the range 2.3-8.2. After five subsequent additions of Co(II) and oxone to the solution, COD and TOC removals increased up to 64% and 33%, respectively. Radical quenching tests confirmed that sulfate radical was the dominant radical species in the system. The main identified by-products from surfactant degradation were: (a) low molecular weight organic acids; (b) aldehydes and formates with shorter ethoxy chain than Brij 35; (c) alcohol ethoxylates carrying hydroxyl groups bonded to ethoxy chain. By-products identification allowed to hypothesize the pathways of Brij 35 degradation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
OXONE®, Monopersulfatverbindung
Sigma-Aldrich
OXONE®, Monopersulfatverbindung, free-flowing, Redi-Dri