- Differentiating intrinsic SERS spectra from a mixture by sampling induced composition gradient and independent component analysis.
Differentiating intrinsic SERS spectra from a mixture by sampling induced composition gradient and independent component analysis.
The Analyst (2011-11-03)
Justin L Abell, Joonsang Lee, Qun Zhao, Harold Szu, Yiping Zhao
PMID22046582
ZUSAMMENFASSUNG
By generating a composition gradient on a highly uniform SERS substrate and applying independent component analysis, we demonstrate that one can extract the intrinsic SERS spectrum of individual components from SERS spectra obtained from a two-component mixture.