Merck
  • Indigenous American ancestry is associated with arsenic methylation efficiency in an admixed population of northwest Mexico.
Indigenous American ancestry is associated with arsenic methylation efficiency in an admixed population of northwest Mexico.
Journal of toxicology and environmental health. Part A (2011-11-04)
Paulina Gomez-Rubio, Yann C Klimentidis, Ernesto Cantu-Soto, Maria M Meza-Montenegro, Dean Billheimer, Zhenqiang Lu, Zhao Chen, Walter T Klimecki
ZUSAMMENFASSUNG
Many studies provide evidence relating lower human arsenic (As) methylation efficiency, represented by high percent urinary monomethylarsonic acid (MMA(V)), with several As-induced diseases, possibly due to the fact that MMA(V) serves as a proxy for MMA(III), the most toxic As metabolite. Some epidemiological studies suggested that indigenous Americans (AME) methylate As more efficiently; however, data supporting this have been equivocal. The aim of this study was to characterize the association between AME ancestry and As methylation efficiency using a panel of ancestry informative genetic markers to determine individual ancestry proportions in an admixed population (composed of two or more isolated ancestral populations) of 746 individuals environmentally exposed to As in northwest Mexico. Total urinary As (TAs) mean and range were 170.4 and 2.3-1053.5 μg/L, while percent AME (%AME) mean and range were 72.4 and 23-100. Adjusted (gender, age, AS3MT 7388/M287T haplotypes, body mass index [BMI], and TAs) multiple regression model showed that higher AME ancestry is significantly associated with lower percentage of urinary As excreted as MMA(V) (%uMMA) in this population (p < .01). Data also demonstrated a significant interaction between BMI and gender, indicating negative association between BMI and %uMMA, stronger in women than men (p < .01). Moreover, age and the AS3MT variants 7388 (intronic) and M287T (nonsynonymous) were also significantly associated with As methylation efficiency (p < .01). This study highlights the importance of BMI and indigenous American ancestry in some of the observed variability in As methylation efficiency, underscoring the need to be considered in epidemiology studies, particularly those carried out in admixed populations.