Merck
A novel solution-stamping process for preparation of a highly conductive aluminum thin film.
Advanced materials (Deerfield Beach, Fla.) (2011-11-05)
Hye Moon Lee, Si-Young Choi, Kyung Tae Kim, Jung-Yeul Yun, Dae Soo Jung, Seung Bin Park, Jongwook Park
ZUSAMMENFASSUNG
A novel solution-stamping process for the preparation of a highly conductive aluminum thin film on both rigid and flexible substrates is proposed. The superior electrical properties of Al thin films fabricated by the solution-stamping process compared to silver and gold films fabricated from colloidal nanoparticles are experimentally demonstrated, and their applications in electronic circuits on rigid and flexible substrates and to organic light-emitting diodes (OLEDs) are investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(ethylenterephthalat), granular
Sigma-Aldrich
Natronkalk, ACS reagent, granular, +100 mesh particle size
Sigma-Aldrich
Natronkalk, p.a., with indicator, pellets