Merck
Functional polymer laminates from hyperthermal hydrogen induced cross-linking.
Langmuir : the ACS journal of surfaces and colloids (2011-11-09)
David B Thompson, Tomas Trebicky, Patrick Crewdson, Matthew J McEachran, Goran Stojcevic, Gilles Arsenault, Woon M Lau, Elizabeth R Gillies
ZUSAMMENFASSUNG
The use of a hyperthermal hydrogen induced cross-linking process to prepare laminates comprising polypropylene, poly(isobutylene-co-isoprene), and poly(vinyl acetate) is described. In this new, milder alternative to conventional plasma techniques, neutral molecular hydrogen projectiles were used to create carbon radicals on impacted surfaces by collision-induced dissociation of C-H bonds, and this process was used to cross-link polymers on a polypropylene surface. It was demonstrated that multiple layers of cross-linked materials could be added, creating polymer laminates with each layer introducing new functionalities and properties. In particular, the present work shows that the process is largely nondestructive toward ester functionalities. First, the esters were grafted to become nonleachable. Then, the esters were subsequently hydrolyzed to convert the surface from hydrophobic to hydrophilic. Afterward, the esters could be recovered by simple esterification demonstrating that further chemical transformations were possible.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylacetat), average Mw ~80,000-120,000 by GPC, beads
Sigma-Aldrich
Polyisobutylen, average Mw ~500,000, average Mn ~200,000 by GPC/MALLS, average Mv ~420,000
Sigma-Aldrich
Polyisobutylen, average Mv ~1,200,000
Sigma-Aldrich
Poly(vinylacetat), average Mw ~500,000 by GPC
Sigma-Aldrich
Kollicoat® SR 30 D, 28.5-31.5% solids basis
Poly(vinylacetat)-Dispersion 30 %, European Pharmacopoeia (EP) Reference Standard