- Volatile organic compound adsorption on a nonporous silica surface: how do different probe molecules sense the same surface?
Volatile organic compound adsorption on a nonporous silica surface: how do different probe molecules sense the same surface?
Langmuir : the ACS journal of surfaces and colloids (2011-11-09)
Carmelo Herdes, M Manuela L Ribeiro Carrott, Patrícia A Russo, Peter J M Carrott
PMID22060183
ZUSAMMENFASSUNG
In this work, we compare experimental results to molecular simulation results of volatile organic compound (VOC) adsorption on nonporous silica. We adopted an effective model for the rough solid surface, obtained by a temperature annealing scheme, plus an experimental/simulation nitrogen adsorption tuning process over the silica energetic oxygen parameter. The measurement/prediction of selected VOCs, specifically, n-pentane and methylcyclohexane, is presented in terms of adsorption isotherms, with an emphasis on the angle distribution analysis of the three studied probe molecules with respect to the same modeled surface.