Merck
[Effect of Limax lyophilized powder on bronchial asthma].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-11-10)
Peng-ke Yan, Guang-qing Lin, Qi-fu Luo, Jin-kui Xie
ZUSAMMENFASSUNG
To study the effect of Limax lyophilized powder on bronchial asthma. The allergic asthma model was established in guinea pigs by combined utilization of aluminum hydroxide and egg albumin to investigate the effect of Limax lyophilized powder on the bronchial flow and on the level of inflammator in bronchoalveolar lavage and serum. The mortality of asthma laboratory guinea pigs was reduced and the incubation period of asthma was extended significantly in Limax lyophilized powder groups. Its antiasthmatic effect was as efficient as the control drug (aminophylline). The leucocyte count was decreased in peripheral blood and the bronchoalveolar lavage fluid. The infiltration of pulmonary tissues eosinophil was also significantly reduced. Further more,the most efficient effects was showed in Limax lyophilized powder at the moderate dosage (63 mg/kg). The bronchial perfusion flow was increased and the level of IL-2 and IL-4 in blood serum and bronchoalveolar lavage fluid was decreased obviously in the aminophylline group and Limax lyophilized powder groups at moderate and high dosage. Limax lyophilized powder could inhibit bronchial asthma evidently.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aminophyllin, ≥98%, powder