Merck
  • In vivo efficacy of a novel liposomal formulation of safingol in the treatment of acute myeloid leukemia.
In vivo efficacy of a novel liposomal formulation of safingol in the treatment of acute myeloid leukemia.
Journal of controlled release : official journal of the Controlled Release Society (2011-11-22)
Kuan-Boone Tan, Leong-Uung Ling, Ralph M Bunte, Wee-Joo Chng, Gigi N C Chiu
ZUSAMMENFASSUNG
Prognosis of patients with acute myeloid leukemia (AML) remains poor despite the use of first-line induction chemotherapy. Therefore, it is imperative to find effective treatment for AML patients. Safingol is a bioactive sphingolipid which has demonstrated promising in vitro anti-leukemic properties; however, translation into clinical use is hampered by its low water solubility and dose-limiting hemolysis. The present study is the first to describe a rationally designed liposome formulation of safingol and demonstrate the anti-leukemic potential using a panel of human AML cell lines and patient samples as well as a human xenograft model in SCID mice. Encapsulation efficiency of safingol into liposomes was approximately 100%, and the release of drug followed square-root-of-time release model. The presence of a transmembrane pH gradient completely abolished the biological activity of liposomal safingol. A positive zeta potential, which influenced cellular accumulation of liposomal safingol, was crucial to the anti-leukemic activity. Liposomal safingol was effective against a wide range of AML subtypes with minimal hemolytic toxicity, and was able to extend the median survival time of the U937-inoculated mice to 31 days as compared to 23 days by free drug. The increase in therapeutic efficacy could be related to the increase in systemic drug exposure as a result of liposome encapsulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
L-threo-Dihydrosphingosine, ≥95% (TLC)