Merck
  • Gas chromatographic determination of cresols in the biological fluids of a non-fatal case of cresol intoxication.
Gas chromatographic determination of cresols in the biological fluids of a non-fatal case of cresol intoxication.
Forensic science international (1990-08-01)
M Yashiki, T Kojima, T Miyazaki, F Chikasue, M Ohtani
ZUSAMMENFASSUNG
A simple and rapid method for analysis of free and conjugated cresols in biological fluids was developed. Prior to and following freeing of the conjugated cresols by acid hydrolysis in a sealed ampoule, free cresols were extracted by Extrelut column extraction, determined by gas chromatography, and confirmed by gas chromatography-mass spectrometry. In a non-fatal case of cresol intoxication a 46-year-old male had ingested about 100 ml of a saponated cresol soap solution. The concentrations of xylenol (2,4- and/or 2,5-dimethylphenol) and p- and m-cresol in the serum sample collected on admission were 15.8 micrograms/g, 43.3 micrograms/g and 73.8 micrograms/g, respectively. The total cresol concentration of 117 micrograms/g in the serum is within the range of fatal concentrations, and it is suspected therefore that the patient's recovery was due to adequate therapy alone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,5-Dimethylphenol, ≥99%
Sigma-Aldrich
3,5-Dimethylphenol, 98%