- Interaction of acetone, hydroxyacetone, acetaldehyde and benzaldehyde with the surface of water ice and HNO3·3H2O ice.
Interaction of acetone, hydroxyacetone, acetaldehyde and benzaldehyde with the surface of water ice and HNO3·3H2O ice.
Physical chemistry chemical physics : PCCP (2011-11-24)
Jérôme Lasne, Carine Laffon, Philippe Parent
PMID22109680
ZUSAMMENFASSUNG
Oxygenated volatile organic compounds (OVOCs) influence the oxidative properties of the atmosphere, and their transport from the ground may occur by scavenging by the HNO(3)-rich supercooled water droplets found in polluted convective air masses. With infrared spectroscopy, we have studied the interactions of four typical atmospheric OVOCs (acetone, hydroxyacetone, acetaldehyde and benzaldehyde) with model surfaces of water ice and of trihydrated nitric acid (NAT) ice. We show that these molecules weakly adsorb on water ice and NAT by hydrogen bonding. No chemical reaction occurs between the molecules and the NAT substrate, the OVOCs remaining intact when in contact with hydrated HNO(3) in atmospheric ice clouds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydroxyaceton, contains ≤500 ppm sodium carbonate as stabilizer, technical grade, 90%