Merck
  • Facile synthesis of hierarchical MoS₂ microspheres composed of few-layered nanosheets and their lithium storage properties.
Facile synthesis of hierarchical MoS₂ microspheres composed of few-layered nanosheets and their lithium storage properties.
Nanoscale (2011-11-26)
Shujiang Ding, Dongyang Zhang, Jun Song Chen, Xiong Wen David Lou
ZUSAMMENFASSUNG
In this work, we demonstrate an interesting polystyrene microsphere-assisted synthesis of hierarchical MoS(2) spheres composed of ultrathin nanosheets. The as-prepared sample exhibits promising lithium storage properties with improved cyclic capacity retention and rate capability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Molybdän(IV)-sulfid, powder
Sigma-Aldrich
Molybdän(IV)-sulfid, powder, <2 μm, 98%