Merck
  • Recombinant cannabinoid type 2 receptor in liposome model activates g protein in response to anionic lipid constituents.
Recombinant cannabinoid type 2 receptor in liposome model activates g protein in response to anionic lipid constituents.
The Journal of biological chemistry (2011-12-03)
Tomohiro Kimura, Alexei A Yeliseev, Krishna Vukoti, Steven D Rhodes, Kejun Cheng, Kenner C Rice, Klaus Gawrisch
ZUSAMMENFASSUNG
Human cannabinoid type 2 (CB(2)) receptor expressed in Escherichia coli was purified and successfully reconstituted in the functional form into lipid bilayers composed of POPC, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (POPS), and cholesteryl hemisuccinate (CHS). Reconstitution was performed by detergent removal from the protein/lipid/detergent mixed micelles either on an adsorbent column, or by rapid dilution to below the critical micelle concentration of detergent followed by removal of detergent monomers on a concentrator. Proteoliposomes prepared at a protein/phospholipid/CHS molar ratio of 1/620-650/210-220 are free of detergent as shown by (1)H NMR, have a homogeneous protein/lipid ratio shown by isopycnic gradient ultracentrifugation, and are small in size with a mean diameter of 150-200 nm as measured by dynamic light scattering. Functional integrity of the reconstituted receptor was confirmed by quantitative binding of (2)H-labeled agonist CP-55,940-d(6) measured by (2)H magic angle spinning NMR, as well as by activation of G protein. The efficiency of G protein activation by agonist-bound CB(2) receptor was affected by negative electric surface potentials of proteoliposomes controlled by the content of anionic CHS or POPS. The activation was highest at an anionic lipid content of about 50 mol %. There was no correlation between the efficiency of G protein activation and an increase of hydrocarbon chain order induced by CHS or cholesterol. The results suggest the importance of anionic lipids in regulating signal transduction by CB(2) receptor and other class A GPCR. The successful reconstitution of milligram quantities of pure, functional CB(2) receptor enables a wide variety of structural studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterylhemisuccinat