Merck
  • Synthesis of highly functionalized chiral 3,3'-disubstituted oxindoles via an organocatalytic enantioselective Michael addition of nitroalkanes to indolylidenecyanoacetates.
Synthesis of highly functionalized chiral 3,3'-disubstituted oxindoles via an organocatalytic enantioselective Michael addition of nitroalkanes to indolylidenecyanoacetates.
Organic letters (2011-12-14)
Lu Liu, Deyan Wu, Shu Zheng, Tengfei Li, Xiangmin Li, Sinan Wang, Jian Li, Hao Li, Wei Wang
ZUSAMMENFASSUNG
An efficient bifunctional cinchona alkaloid derived thiourea-promoted enantioselective conjugate addition of nitroalkanes to indolylidenecyanoacetates has been developed under neat conditions. The process leads to synthetically interesting densely functionalized 3,3'-disubstituted oxindoles with creation of up to three stereogenic centers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyanessigsäure, 99%