Merck
Occupational safety of farmers in the vegetable industry.
International journal of occupational safety and ergonomics : JOSE (2011-12-14)
Jinky Leilanie D P Lu
ZUSAMMENFASSUNG
This study aimed to determine unsafe farming practices, nonergonomically designed equipment, and possible adverse health effects of pesticide exposure. Data were gathered with a structured survey, physical examination and blood extraction. Environmental monitoring was done for vegetables to determine multipesticide residue. Forty-eight percent of respondents reported feeling sick because of work. Vegetable residue was found in 16.67% of the samples. Vegetable profenofos of appoximately1 mg exceeded the necessary average daily intake of a maximum of 0.006 mg. Data also showed nonergonomically designed tools and unsafe work practices that increased pesticide exposure such as re-entering recently sprayed areas, a damaged backpack sprayer, spills during spraying, and no ventilation in greenhouses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Profenofos, PESTANAL®, analytical standard