Merck
  • A catalytic version of hypervalent aryl-λ3-iodane-induced Hofmann rearrangement of primary carboxamides: iodobenzene as an organocatalyst and m-chloroperbenzoic acid as a terminal oxidant.
A catalytic version of hypervalent aryl-λ3-iodane-induced Hofmann rearrangement of primary carboxamides: iodobenzene as an organocatalyst and m-chloroperbenzoic acid as a terminal oxidant.
Chemical communications (Cambridge, England) (2011-12-14)
Kazunori Miyamoto, Yuuta Sakai, Shunsuke Goda, Masahito Ochiai
ZUSAMMENFASSUNG
The first catalytic version of hypervalent aryl-λ(3)-iodane-induced Hofmann rearrangement of primary carboxamides, which probably involves in situ generation of a tetracoordinated bis(aqua)(hydroxy)phenyl-λ(3)-iodane complex as an active oxidant from a catalytic amount of iodobenzene by the reaction with m-chloroperbenzoic acid in the presence of HBF(4) in dichloromethane-water under mild conditions, was developed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Chlorperbenzoesäure, ≤77%
Sigma-Aldrich
Iodbenzol, 98%