Merck
Low frequency sonophoresis mediated transdermal and intradermal delivery of ketoprofen.
International journal of pharmaceutics (2011-12-17)
Anushree Herwadkar, Vishal Sachdeva, Leslie F Taylor, Herb Silver, Ajay K Banga
ZUSAMMENFASSUNG
The objective of this study was to test low frequency sonophoresis at 20 kHz for delivery of ketoprofen into and across the skin. Permeation studies were carried out in vitro on excised hairless rat skin over a period of 24h using Franz diffusion cells after which, skin samples were subjected to skin extraction to quantify the amount of drug present in skin. Parameters like ultrasound application time, duty cycle coupling medium and distance of ultrasound horn from skin were optimized. Transepidermal water loss (TEWL) was measured to indicate the extent of barrier disruption following sonophoresis. Confocal microscopy was used to visualize dye penetration through sonophoresis treated skin. Application of ultrasound significantly enhanced permeation of ketoprofen from 74.87 ± 5.27 μg/cm(2) for passive delivery to 491.37 ± 48.78 μg/cm(2) for sonophoresis. Drug levels in skin layers increased from 34.69 ± 7.25 μg following passive permeation to 212.62 ± 45.69 μg following sonophoresis. TEWL increased from 31.6 ± 0.02 (passive) to 69.5 ± 12.60 (sonophoresis) indicating disruption of barrier properties. Confocal microscopy images depicted enhanced dye penetration through sonophoresis treated skin confirming barrier disruption. Low frequency sonophoresis with optimized ultrasound parameters can be effectively used to actively enhance transdermal and topical delivery of ketoprofen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumlaurylsulfat, BioReagent, Molecular Biology, suitable for electrophoresis
Sigma-Aldrich
Ammoniumlaurylsulfat -Lösung, ~30% in H2O
Sigma-Aldrich
Lithiumlaurylsulfat, ≥98.5% (GC)
Sigma-Aldrich
Lithiumlaurylsulfat, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Ammoniumlaurylsulfat -Lösung, ~30% in H2O (T)