Merck
Sulfadimethoxine and sulfaguanidine: their sorption potential on natural soils.
Chemosphere (2011-12-27)
Anna Białk-Bielińska, Joanna Maszkowska, Wojciech Mrozik, Agata Bielawska, Marta Kołodziejska, Richard Palavinskas, Piotr Stepnowski, Jolanta Kumirska
ZUSAMMENFASSUNG
Sulfonamides (SAs) are one of the oldest groups of veterinary chemotherapeutic agents. As these compounds are not completely metabolized in animals, a high proportion of the native form is excreted in feces and urine. They are therefore released either directly to the environment in aquacultures and by grazing animals, or indirectly during the application of manure or slurry. Once released into the environment, SAs become distributed among various environmental compartments and may be transported to surface or ground waters. The physicochemical properties of SAs, dosage and nature of the matrix are the factors mainly responsible for their distribution in the natural environment. Although these rather polar compounds have been in use for over half a century, knowledge of their fate and behavior in soil ecosystems is still limited. Therefore, in this work we have determined the sorption potential of sulfadimethoxine and sulfaguanidine on various natural soils. The influence on sorption of external factors, such as ionic strength and pH, were also determined. The sorption coefficients (K(d)) obtained for the sulfonamides investigated were quite low (from 0.20 to 381.17 mL g(-1) for sulfadimethoxine and from 0.39 to 35.09 mL g(-1) for sulfaguanidine), which indicated that these substances are highly mobile and have the potential to run off into surface waters and/or infiltrate ground water. Moreover, the sorption of these pharmaceuticals was found to be influenced by OC, soil solution pH and ionic strength, with higher K(d) values for soils of higher OC and lower K(d) values with increasing pH and ionic strength.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulfaguanidin
Supelco
Sulfadimethoxin, VETRANAL®, analytical standard
Supelco
Sulfaguanidin, VETRANAL®, analytical standard
Supelco
Sulfadimethoxin, 98.0-102.0%