Merck
Purification of egg yolk phosvitin by anion exchange chromatography.
Journal of chromatography. A (2011-12-27)
Bo Lei, Jianping Wu
ZUSAMMENFASSUNG
The objective of this study was to develop a simple method of phosvitin purification from hen egg yolk without using organic solvents. Egg yolk was diluted with equal volume of water and stirred for one hour at room temperature, followed by centrifugation to remove soluble proteins along with most of the yolk lipids in the supernatant. The granules were collected as the precipitate containing minimum amount of lipids (dry granules). The dry granules were dissolved in 0.05 M carbonate-bicarbonate buffer at pH 9.6, which yields a light yellowish solution used for anion exchange chromatography. Phosvitin fraction was collected from anion exchange chromatography as the last eluting peak with a purity of 92.6% and a yield of 35.4% of total phosvitin in the yolk or a recovery of 1.9% of total yolk dry matter, which are comparable to current methods employing organic solvents or chromatography after salt fractionation and dialysis. This method developed is simple and fast without using organic solvents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosvitin aus Eigelb