Merck
  • TTC-based screening assay for ω-transaminases: a rapid method to detect reduction of 2-hydroxy ketones.
TTC-based screening assay for ω-transaminases: a rapid method to detect reduction of 2-hydroxy ketones.
Journal of biotechnology (2012-01-10)
Torsten Sehl, Robert C Simon, Helen C Hailes, John M Ward, Ursula Schell, Martina Pohl, Dörte Rother
ZUSAMMENFASSUNG
A rapid TTC-based screening assay for ω-transaminases was developed to determine the conversion of substrates with a 2-hydroxy ketone motif. Oxidation of the compounds in the presence of 2,3,5-triphenyltetrazolium chloride (TTC) results in a reduction of the colourless TTC to a red-coloured 1,3,5-triphenylformazan. The enzymatic reductive amination of a wide range of various aliphatic, aliphatic-aromatic and aromatic-aromatic 2-hydroxy ketones can be determined by the decrease of the red colouration due to substrate consumption. The conversion can be quantified spectrophotometrically at 510 nm based on reactions, e.g. with crude cell extracts in 96-well plates. Since the assay is independent of the choice of diverse amine donors a panel of ω-transaminases was screened to detect conversion of 2-hydroxy ketones with three different amine donors: l-alanine, (S)-α-methylbenzylamine and benzylamine. The results could be validated using HPLC and GC analyses, showing a deviation of only 5-10%. Using this approach enzymes were identified demonstrating high conversions of acetoin and phenylacetylcarbinol to the corresponding amines. Among these enzymes three novel wild-type ω-transaminases have been identified.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,3,5-Triphenyltetrazoliumchlorid, ≥98.0% (HPLC)
Millipore
2,3,5-Triphenyl-tetrazoliumchlorid -Lösung, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC