Merck
  • Tetraethylorthosilicate as molecular precursor to the formation of amorphous silica networks. A DFT-SCRF study of the base catalyzed hydrolysis.
Tetraethylorthosilicate as molecular precursor to the formation of amorphous silica networks. A DFT-SCRF study of the base catalyzed hydrolysis.
Journal of molecular modeling (2012-01-19)
Lorenzo Fernandez, Iñaki Tuñón, Julio Latorre, Carmen Guillem, Aurelio Beltrán, Pedro Amorós
ZUSAMMENFASSUNG
Quantum chemical calculations using density functional theory have been carried out to investigate two chemical pathways for the last step of the hydrolysis of tetraethylorthosilicate (TEOS) in basic catalyzed environment. The two models that are introduced in this study depend on the number of water molecules involved at the base catalyzed hydrolysis. Solution equilibrium geometries of the molecules involved in the transition states, reactants and product complexes of the two chemical pathways were fully optimized at B3LYP level of theory with the standard 6-31+G(d) basis set, modeling solvent effects using a polarizable continuum solvation model (PCM). Both models predict relative low activation energies. However, the model with two water molecules seems to be more adequate to describe the basic hydrolysis. A natural bond orbital (NBO) analysis seems to show that the proton transfer from water to ethoxy group would occur through a large hyperconjugative interaction, LP(O) → σ*(O-H), which is related to the nonbonding oxygen lone pair orbital from ethoxy group with the vicinal σ*(O-H) anti bonding orbital O-H of a water molecule.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems