Merck
  • Effect of long-term zinc pollution on soil microbial community resistance to repeated contamination.
Effect of long-term zinc pollution on soil microbial community resistance to repeated contamination.
Bulletin of environmental contamination and toxicology (2012-01-27)
Beata Klimek
ZUSAMMENFASSUNG
The aim of the study was to compare the effects of stress (contamination trials) on the microorganisms in zinc-polluted soil (5,018 mg Zn kg(-1) soil dry weight) and unpolluted soil (141 mg Zn kg(-1) soil dw), measured as soil respiration rate. In the laboratory, soils were subjected to copper contamination (0, 500, 1,500 and 4,500 mg kg(-1) soil dw), and then a bactericide (oxytetracycline) combined with a fungicide (captan) along with glucose (10 mg g(-1) soil dw each) were added. There was a highly significant effect of soil type, copper treatment and oxytetracycline/captan treatment. The initial respiration rate of chronically zinc-polluted soil was higher than that of unpolluted soil, but in the copper treatment it showed a greater decline. Microorganisms in copper-treated soil were more susceptible to oxytetracycline/captan contamination. After the successive soil contamination trials the decline of soil respiration was greater in zinc-polluted soil than in unpolluted soil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Captan, PESTANAL®, analytical standard