Merck
  • Chemical generation of o-quinone monoimines for the rapid construction of 1,4-benzoxazine derivatives.
Chemical generation of o-quinone monoimines for the rapid construction of 1,4-benzoxazine derivatives.
Organic & biomolecular chemistry (2012-01-31)
Naganjaneyulu Bodipati, Rama Krishna Peddinti
ZUSAMMENFASSUNG
Highly reactive o-benzoquinone monoimines were chemically generated and successfully trapped with electron-rich olefins that led to the synthesis of hitherto unknown 1,4-benzoxazine derivatives. This unprecedented transformation was achieved by the oxidation of o-aminophenols bearing appropriate functionality on the arene residue with less expensive hypervalent iodine reagent in the presence of vinylic ethers or thioethers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminophenol, 99%