Merck
Basic principles in preclinical cancer chemotherapy.
Journal of cancer research and clinical oncology (1990-01-01)
N Brock, J Pohl, B Schneider
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bis-(2-chlorethyl)-amin -hydrochlorid, 98%