- Effects of a lactobacilli, oligosaccharide and organic germanium intake on the immune responses of mice.
Effects of a lactobacilli, oligosaccharide and organic germanium intake on the immune responses of mice.
Bioscience, biotechnology, and biochemistry (2012-02-09)
Takashi Nakamura, Miki Saito, Hisashi Aso
PMID22313762
ZUSAMMENFASSUNG
The organic germanium compound, Ge-132, has immune-modulating effects. We evaluated the symbiotic effects of Ge-132 with lactobacilli and oligosaccharide (LB/OS) on the immune responses of mice. The highest fecal IgA levels were observed in the mice receiving a low concentration of Ge-132 with LB/OS for 8 weeks. Our data suggest that LB/OS with a low concentration of Ge-132 stimulated the intestinal immunity.