Merck
Uranium(VI) reduction by iron(II) monosulfide mackinawite.
Environmental science & technology (2012-02-10)
Sung Pil Hyun, James A Davis, Kai Sun, Kim F Hayes
ZUSAMMENFASSUNG
Reaction of aqueous uranium(VI) with iron(II) monosulfide mackinawite in an O(2) and CO(2) free model system was studied by batch uptake measurements, equilibrium modeling, and L(III) edge U X-ray absorption spectroscopy (XAS). Batch uptake measurements showed that U(VI) removal was almost complete over the wide pH range between 5 and 11 at the initial U(VI) concentration of 5 × 10(-5) M. Extraction by a carbonate/bicarbonate solution indicated that most of the U(VI) removed from solution was reduced to nonextractable U(IV). Equilibrium modeling using Visual MINTEQ suggested that U was in equilibrium with uraninite under the experimental conditions. X-ray absorption near edge structure (XANES) and extended X-ray absorption fine structure (EXAFS) spectroscopy showed that the U(IV) phase associated with mackinawite was uraninite. Oxidation experiments with dissolved O(2) were performed by injecting air into the sealed reaction bottles containing mackinawite samples reacted with U(VI). Dissolved U measurement and XAS confirmed that the uraninite formed from the U(VI) reduction by mackinawite did not oxidize or dissolve under the experimental conditions. This study shows that redox reactions between U(VI) and mackinawite may occur to a significant extent, implying an important role of the ferrous sulfide mineral in the redox cycling of U under sulfate reducing conditions. This study also shows that the presence of mackinawite protects uraninite from oxidation by dissolved O(2). The findings of this study suggest that uraninite formation by abiotic reduction by the iron sulfide mineral under low temperature conditions is an important process in the redistribution and sequestration of U in the subsurface environments at U contaminated sites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eisen(II)-sulfid, −100 mesh, 99.9% trace metals basis