Merck
  • Pd2(dba)3/P(MeNCH2CH2)3N·HCl-mediated Stille cross-coupling of [1-11C]acetyl chloride with aryl and heteroaryl stannanes.
Pd2(dba)3/P(MeNCH2CH2)3N·HCl-mediated Stille cross-coupling of [1-11C]acetyl chloride with aryl and heteroaryl stannanes.
Nuclear medicine and biology (2012-02-18)
Takuya Arai
ZUSAMMENFASSUNG
Developments in the Stille cross-coupling of [1-11C]acetyl chloride with tributylphenylstannane mediated by the Pd2(dba)3/P(MeNCH2CH2)3N·HClsystem are reported. The reaction conditions for the cross-coupling of [1-11C]acetyl chloride with tributylphenylstannane were optimized, and the reaction scope was investigated. The cross-couplings of [1-11C]acetyl chloride with a range of aryl and heteroaryl stannanes were performed with good to excellent radiochemical conversions using the developed method. A highly efficient method for the Stille cross-coupling of [1-11C]acetyl chloride with organostannanes was demonstrated. It is anticipated that our method will be an attractive addition to the carbon-11-labeling procedures available for the synthesis of positron emission tomography probes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetylchlorid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetylchlorid, reagent grade, 98%
Sigma-Aldrich
Acetylchlorid, reagent grade, 98%