Merck
  • Akt inhibitors MK-2206 and nelfinavir overcome mTOR inhibitor resistance in diffuse large B-cell lymphoma.
Akt inhibitors MK-2206 and nelfinavir overcome mTOR inhibitor resistance in diffuse large B-cell lymphoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-02-18)
Adam M Petrich, Violetta Leshchenko, Pei-Yu Kuo, Bing Xia, Venu K Thirukonda, Netha Ulahannan, Shanisha Gordon, Melissa J Fazzari, B Hilda Ye, Joseph A Sparano, Samir Parekh
ZUSAMMENFASSUNG
The mTOR pathway is constitutively activated in diffuse large B-cell lymphoma (DLBCL). mTOR inhibitors have activity in DLBCL, although response rates remain low. We evaluated DLBCL cell lines with differential resistance to the mTOR inhibitor rapamycin: (i) to identify gene expression profile(s) (GEP) associated with resistance to rapamycin, (ii) to understand mechanisms of rapamycin resistance, and (iii) to identify compounds likely to synergize with mTOR inhibitor. We sought to identify a GEP of mTOR inhibitor resistance by stratification of eight DLBCL cell lines with respect to response to rapamycin. Then, using pathway analysis and connectivity mapping, we sought targets likely accounting for this resistance and compounds likely to overcome it. We then evaluated two compounds thus identified for their potential to synergize with rapamycin in DLBCL and confirmed mechanisms of activity with standard immunoassays. We identified a GEP capable of reliably distinguishing rapamycin-resistant from rapamycin-sensitive DLBCL cell lines. Pathway analysis identified Akt as central to the differentially expressed gene network. Connectivity mapping identified compounds targeting Akt as having a high likelihood of reversing the GEP associated with mTOR inhibitor resistance. Nelfinavir and MK-2206, chosen for their Akt-inhibitory properties, yielded synergistic inhibition of cell viability in combination with rapamycin in DLBCL cell lines, and potently inhibited phosphorylation of Akt and downstream targets of activated mTOR. GEP identifies DLBCL subsets resistant to mTOR inhibitor therapy. Combined targeting of mTOR and Akt suppresses activation of key components of the Akt/mTOR pathway and results in synergistic cytotoxicity. These findings are readily adaptable to clinical trials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder