Merck
Electrophysiological effects of hydrogen sulfide on human atrial fibers.
Chinese medical journal (2012-02-22)
Meng Xu, Yu-Ming Wu, Qian Li, Su Liu, Qian Li, Rui-Rong He
ZUSAMMENFASSUNG
It has been reported that endogenous or exogenous hydrogen sulfide (H(2)S) exerts physiological effects in the vertebrate cardiovascular system. We have also demonstrated that H(2)S acts as an important regulator of electrophysiological properties in guinea pig papillary muscles and on pacemaker cells in sinoatrial nodes of rabbits. This study was to observe the electrophysiological effects of H(2)S on human atrial fibers. Human atrial samples were collected during cardiac surgery. Parameters of action potential in human atrial specialized fibers were recorded using a standard intracellular microelectrode technique. NaHS (H(2)S donor) (50, 100 and 200 µmol/L) decreased the amplitude of action potential (APA), maximal rate of depolarization (V(max)), velocity of diastolic (phase 4) depolarization (VDD) and rate of pacemaker firing (RPF), and shortened the duration of 90% repolarization (APD(90)) in a concentration-dependent manner. ATP-sensitive K(+) (K(ATP)) channel blocker glibenclamide (Gli, 20 µmol/L) partially blocked the effects of NaHS (100 µmol/L) on human atrial fiber cells. The L-type Ca(2+) channel agonist Bay K8644 (0.5 µmol/L) also partially blocked the effects of NaHS (100 µmol/L). An inhibitor of cystathionine γ-lyase (CSE), DL-propargylglycine (PPG, 200 µmol/L), increased APA, V(max), VDD and RPF, and prolonged APD(90). H(2)S exerts a negative chronotropic action and accelerates the repolarization of human atrial specialized fibers, possibly as a result of increases in potassium efflux through the opening of K(ATP) channels and a concomitant decrease in calcium influx. Endogenous H(2)S may be generated by CSE and act as an important regulator of electrophysiological properties in human atrial fibers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid