Merck
  • Total synthesis of flocoumafen via knoevenagel condensation and intramolecular ring cyclization: general access to natural products.
Total synthesis of flocoumafen via knoevenagel condensation and intramolecular ring cyclization: general access to natural products.
Molecules (Basel, Switzerland) (2012-02-23)
Jae-Chul Jung, Eunyoung Lim, Yongnam Lee, Dongguk Min, Jeremy Ricci, Oe-Sook Park, Mankil Jung
ZUSAMMENFASSUNG
The total synthesis and structure determination of cis- and trans-flocoumafen was described. The key synthetic steps involve Knoevenagel condensation with p-methoxybenzaldehyde, in situ decarboxylation and intramolecular ring cyclization to construct the tetralone skeleton. Stereospecific reduction of the O-alkylated ketone 13 afforded good yield of precusor alcohol 5. Final coupling of alcohol 5 with 4-hydroxy-coumarin yielded flocoumafen (1). Separation and structure determination of cis- and trans-flocoumafen through 2D NMR analyses-assisted computer simulation techniques for the evaluation of anticoagulant activities are reported for the first time. This method is useful for generating the core tetralone skeleton of 4-hydroxycoumarin derivatives and provides a generalized access to various warfarin type anticoagulants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
p-Anisaldehyd, 98%
Sigma-Aldrich
4-Hydroxycumarin, 98%
Supelco
p-Anisaldehyd, analytical standard
Sigma-Aldrich
p-Anisaldehyd, ≥97.5%, FCC, FG
Supelco
Flocoumafen, PESTANAL®, analytical standard