Merck
  • Asymmetric synthesis of (R)-1,3-butanediol from 4-hydroxy-2-butanone by a newly isolated strain Candida krusei ZJB-09162.
Asymmetric synthesis of (R)-1,3-butanediol from 4-hydroxy-2-butanone by a newly isolated strain Candida krusei ZJB-09162.
Applied microbiology and biotechnology (2012-03-01)
Ren-Chao Zheng, Zheng Ge, Zhao-Kuan Qiu, Yuan-Shan Wang, Yu-Guo Zheng
ZUSAMMENFASSUNG
Biocatalytic asymmetric preparation of (R)-1,3-butanediol has been attracting much attention in pharmaceuticals industry. A new ideal strain, ZJB-09162, which exhibited high reduction activity and excellent (R)-stereospecificity towards 4-hydroxy-2-butanone, has been successfully isolated from soil samples. Based on morphology, physiological tests (API 20 C AUX), and 5.8S-ITS sequence, the isolate was identified as Candida krusei. Kinetic characterization demonstrated that carbonyl reductase from C. krusei ZJB-09162 preferred NADH to NADPH as cofactor, indicating it might be a new carbonyl reductase. (R)-1,3-Butanediol was produced in 19.8 g/L, 96.6% conversion, and 99.0% ee at optimal pH 8.5, 35 °C with a 2:1 molar ratio of glucose to 4H2B. In order to achieve higher product titer, the substrate loading was optimized in fixed catalysts and fixed substrate/catalysts ratio mode. The bioreduction of 4-hydroxy-2-butanone at a concentration of 45.0 g/L gave (R)-1,3-butanediol in 38.7 g/L and 83.9% conversion. Therefore, C. krusei ZJB-09162 was, for the first time, proven to be a promising biocatalyst for enzymatic preparation of (R)-1,3-butanediol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanon, FCC, FG
Sigma-Aldrich
(±)-1,3-Butandiol, ReagentPlus®, 99.5%
Supelco
2-Butanon, analytical standard
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
(±)-1,3-Butandiol, anhydrous, ≥99%
Sigma-Aldrich
(R)-(−)-1,3-Butandiol, 95%